4-Fluorostyrene, 97%, stabilized
4-Fluorostyrene, 97%, stabilized
4-Fluorostyrene, 97%, stabilized
Thermo Scientific Chemicals

4-Fluorostyrene, 97%, stabilized

CAS: 405-99-2 | C8H7F | 122.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 168340010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS405-99-2
IUPAC Name1-ethenyl-4-fluorobenzene
Molecular FormulaC8H7F
InChI KeyJWVTWJNGILGLAT-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Refractive index1.5140 to 1.5160 (20°C, 589 nm)
Stabilizer0.1 % tert.-butylcatechol
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only