2-Methyltetrahydrofuran, 99+%, pure, stabilized
2-Methyltetrahydrofuran, 99+%, pure, stabilized
2-Methyltetrahydrofuran, 99+%, pure, stabilized
Thermo Scientific Chemicals

2-Methyltetrahydrofuran, 99+%, pure, stabilized

CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Metal Drum
Catalog number 168360100
Price (USD)
812.65
Special offer
Online exclusive
Ends: 28-Dec-2026
956.00
Save 143.35
Each
Quantity:
10 kg
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)
812.65
Special offer
Online exclusive
Ends: 28-Dec-2026
956.00
Save 143.35
Each
Chemical Identifiers
CAS96-47-9
IUPAC Name2-methyloxolane
Molecular FormulaC5H10O
InChI KeyJWUJQDFVADABEY-UHFFFAOYNA-N
SMILESCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Water=<0.2 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only