Choose the new Thermo Scientific Nunc EasYDish cell culture dishes for improved handling and stacking.
Nunc™ Cell Culture/Petri Dishes
Actual product may vary
Nunc™ Cell Culture/Petri Dishes
Nunc™ Cell Culture/Petri Dishes
Thermo Scientific™

Nunc™ Cell Culture/Petri Dishes

Perform a variety of applications with cell culture/Petri dishes offered in a broad range of sizes and styles to suit a variety of experimental needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture AreaSurface TreatmentGriddedVent
168381145 sq. cmNunclon DeltaNot GriddedLid with airvent
1503188.8 sq. cmNunclon DeltaNot GriddedLid without airvent
1530668.8 sq. cmNunclon DeltaNot GriddedLid with airvent
1749268.8 sq. cmNunclon DeltaGriddedLid with airvent
1710998.8 sq. cmUntreatedNot GriddedLid with airvent
17488820.8 sq. cmCell CultureNot GriddedLid with airvent
15032621.5 sq. cmNunclon DeltaNot GriddedLid without airvent
15028821.5 sq. cmNunclon DeltaNot GriddedLid with airvent
16955821.5 sq. cmNunclon DeltaGriddedLid with airvent
15035056.7 sq. cmNunclon DeltaNot GriddedLid with airvent
17293156.7 sq. cmNunclon DeltaNot GriddedLid with airvent
Catalog number 168381
Price (USD)
420.65
Online exclusive
457.00
Save 36.35
Case of 80
Culture Area:
145 sq. cm
Surface Treatment:
Nunclon Delta
Gridded:
Not Gridded
Vent:
Lid with airvent
Price (USD)
420.65
Online exclusive
457.00
Save 36.35
Case of 80

With wide selections in size and style, Thermo Scientific Nunc cell culture dishes suit a multitude of experimental needs in your cell culture routine. Perform a variety of applications in dishes with the Nunclon™ Delta surface treatment that promotes maximum adhesion for a broad range of cell types.

Features

  • Top and bottom stacking rings for easier handling and more stable stacking
  • Sterile and non-pyrogenic
  • Available with or without air vents for gas exchange
  • 2 x 2 mm grid for cloning experiments and determination of plating efficiency

Includes: a certificate that assures user of biological performance as well as traceability

Warranty: 90 days

Additional information

  • Cat. Nos. 174926 and 169558 feature a 2 x 2 mm grid
  • Cat. No. 171099 is for suspension cell culture; it is not Nunclon Delta treated
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure IncludedClosure Included
Closure MaterialPolystyrene
GriddedNot Gridded
IncludesAirvent
No. per Case80/Cs.
No. per Pack10 Pk.
PackagingResealable Bags
Quantity10 Pk., 80/Cs.
ShapeRound
TypeCell Culture Dish
VentLid with airvent
Volume (Metric) Working35 mL
Bottom ShapeFlat
Capacity (Metric)35 mL (Well)
ColorClear
Culture Area145 sq. cm
Diameter (Metric)150 mm
Height (Metric)21 mm
MaterialPolystyrene
SterilitySterile
Surface TreatmentNunclon Delta
Well Count1
Unit SizeCase of 80

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

What is Nunclon Delta?

Nunclon Delta refers to our performance-certified surface for culture of adherent cells, available in a variety of common formats (flasks, plates, multiwell dishes). It is tested with at least four cell lines (varies by product and manufacturing location) commonly used in research, and it should work well for most robust cell lines. It does not involve addition of coating reagent, but rather is manufactured by exposure of the molded products to a proprietary energy source, which renders the surface more hydrophilic by means of increasing the concentration of oxygen-containing (e.g. hydroxyl) molecular groups. It is by this hydrophilic property that anchorage-dependent cells are able to attach to the surface and become confluent.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.