2-Methoxyethanol, 99+%, for spectroscopy
2-Methoxyethanol, 99+%, for spectroscopy
2-Methoxyethanol, 99+%, for spectroscopy
Thermo Scientific Chemicals

2-Methoxyethanol, 99+%, for spectroscopy

CAS: 109-86-4 | C3H8O2 | 76.09 g/mol
数量:
1 L
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 168580025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
さらに表示
Color scale=<10 APHA
Water=<0.1 %
UV1 cm cell vs water
UVat 240 nm A: =<0.3
UVat 250 nm A: =<0.2
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only