Sodium ethoxide, 96%, pure
Sodium ethoxide, 96%, pure
Sodium ethoxide, 96%, pure
Thermo Scientific Chemicals

Sodium ethoxide, 96%, pure

CAS: 141-52-6 | C2H5NaO | 68.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 168595000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS141-52-6
IUPAC Namesodium ethanolate
Molecular FormulaC2H5NaO
InChI KeyQDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES[Na+].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=95 % (NaOC2H5)
Titration with HCl>=98 % (total NaOC2H5)
NaOH + Na2CO3 (as NaOH)=<2 %
Appearance (Color)White to yellow
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only