2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%

CAS: 6976-37-0 | C8H19NO5 | 209.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 168870025
Price (USD)
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS6976-37-0
IUPAC Name2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC8H19NO5
InChI KeyOWMVSZAMULFTJU-UHFFFAOYSA-N
SMILESOCCN(CCO)C(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<1.0 % (K.F.)
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only