Lauroyl chloride, 98%
Lauroyl chloride, 98%
Lauroyl chloride, 98%
Thermo Scientific Chemicals

Lauroyl chloride, 98%

CAS: 112-16-3 | C12H23ClO | 218.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 169032500
Price (USD)
86.40
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
86.40
Each
Chemical Identifiers
CAS112-16-3
IUPAC Namedodecanoyl chloride
Molecular FormulaC12H23ClO
InChI KeyNQGIJDNPUZEBRU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear liquid
GC>=97.5 %
Color scale=<60 APHA
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only