2-Benzoxazolinone, 98%
2-Benzoxazolinone, 98%
2-Benzoxazolinone, 98%
Thermo Scientific Chemicals

2-Benzoxazolinone, 98%

CAS: 59-49-4 | C7H5NO2 | 135.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 169080250
Price (USD)
51.65
Special offer
Online exclusive
Ends: 28-Dec-2026
60.10
Save 8.45
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 28-Dec-2026
60.10
Save 8.45
Each
Chemical Identifiers
CAS59-49-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)White to beige to grey-brown
Infrared spectrumConforms
Melting point137°C to 142°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only