3-Cyanobenzoic acid, 98%
3-Cyanobenzoic acid, 98%
3-Cyanobenzoic acid, 98%
Thermo Scientific Chemicals

3-Cyanobenzoic acid, 98%

CAS: 1877-72-1 | C8H5NO2 | 147.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 169130500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1877-72-1
IUPAC Name3-cyanobenzoic acid
Molecular FormulaC8H5NO2
InChI KeyGYLKKXHEIIFTJH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder
Melting point217°C to 222°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only