Phenanthridine, 98%
Phenanthridine, 98%
Phenanthridine, 98%
Thermo Scientific Chemicals

Phenanthridine, 98%

CAS: 229-87-8 | C13H9N | 179.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 169250010
Price (USD)
43.65
Online exclusive
47.90
Save 4.25
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
43.65
Online exclusive
47.90
Save 4.25
Each
Chemical Identifiers
CAS229-87-8
IUPAC Namephenanthridine
Molecular FormulaC13H9N
InChI KeyRDOWQLZANAYVLL-UHFFFAOYSA-N
SMILESC1=CC=C2C(=C1)C=NC1=CC=CC=C21
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point104°C to 108°C
Appearance (Color)White to beige
GC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only