Iron(III) chloride, 98%, pure, anhydrous
Iron(III) chloride, 98%, pure, anhydrous
Iron(III) chloride, 98%, pure, anhydrous
Thermo Scientific Chemicals

Iron(III) chloride, 98%, pure, anhydrous

CAS: 7705-08-0 | Cl3Fe | 162.20 g/mol
数量:
2.5 kg
10 kg
25 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 169430100
価格(JPY)
-
数量:
10 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7705-08-0
IUPAC Nameiron(3+) trichloride
Molecular FormulaCl3Fe
InChI KeyRBTARNINKXHZNM-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Fe+3]
さらに表示
Appearance (Form)Powder
Manganese (Mn)=<3000 ppm
Arsenic (As)=<20 ppm
Zinc (Zn)=<1000 ppm
Copper (Cu)=<1000 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only