1,3-Dibromo-5,5-dimethylhydantoin, 98%
1,3-Dibromo-5,5-dimethylhydantoin, 98%
1,3-Dibromo-5,5-dimethylhydantoin, 98%
Thermo Scientific Chemicals

1,3-Dibromo-5,5-dimethylhydantoin, 98%

CAS: 77-48-5 | C5H6Br2N2O2 | 285.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 169540025
Price (USD)
244.65
Special offer
Online exclusive
Ends: 28-Dec-2026
288.00
Save 43.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
244.65
Special offer
Online exclusive
Ends: 28-Dec-2026
288.00
Save 43.35
Each
Chemical Identifiers
CAS77-48-5
IUPAC Name1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione
Molecular FormulaC5H6Br2N2O2
InChI KeyVRLDVERQJMEPIF-UHFFFAOYSA-N
SMILESCC1(C)N(Br)C(=O)N(Br)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Titration Iodometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only