Methyl bromoacetate, 99%
Methyl bromoacetate, 99%
Methyl bromoacetate, 99%
Thermo Scientific Chemicals

Methyl bromoacetate, 99%

CAS: 96-32-2 | C3H5BrO2 | 152.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 169550025
Price (USD)
1,176.00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
1,176.00
Each
Chemical Identifiers
CAS96-32-2
IUPAC Namemethyl 2-bromoacetate
Molecular FormulaC3H5BrO2
InChI KeyYDCHPLOFQATIDS-UHFFFAOYSA-N
SMILESCOC(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4570 to 1.4590 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only