Trifluoromethanesulfonic acid, 99%, extra pure
Trifluoromethanesulfonic acid, 99%, extra pure
Trifluoromethanesulfonic acid, 99%, extra pure
Thermo Scientific Chemicals

Trifluoromethanesulfonic acid, 99%, extra pure

Trifluoromethanesulfonic acid, 99%, CHF3O3S, CAS Number-1493-13-6, hotf, methanesulfonic acid, trifluoro, tfmsa, perfluoromethanesulfonic acid, tfoh, trifluoromethanesulphonic acid, triflic acid, trifluoromethane sulfonic acid, trimsylate, trifluoromethylsulfonic acid, 250mL, 03,IV,34, 162.0 deg.C | CAS: 1493-13-6 | CHF3O3S | 150.07 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 mL
10 mL
1 L
Packaging:
Glass bottle
產品號碼 169890011
價格 (TWD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS1493-13-6
IUPAC Nametrifluoromethanesulfonic acid
Molecular FormulaCHF3O3S
InChI KeyITMCEJHCFYSIIV-UHFFFAOYSA-N
SMILESOS(=O)(=O)C(F)(F)F
檢視更多
規格Specification Sheet規格表
Titration with NaOH98.5 to 101.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.3260 to 1.3300 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only