Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tributyl phosphate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 169930010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS126-73-8
IUPAC Nametributyl phosphate
Molecular FormulaC12H27O4P
InChI KeySTCOOQWBFONSKY-UHFFFAOYSA-N
SMILESCCCCOP(=O)(OCCCC)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
Color scale=<50 APHA
Refractive index1.4230 to 1.4250 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only