Rat IgG2a kappa Isotype Control (eBR2a), APC, eBioscience™ - FAQs
Rat IgG2a kappa Isotype Control (eBR2a), APC, eBioscience™ - FAQs
View additional product information for Rat IgG2a kappa Isotype Control (eBR2a), APC, eBioscience™ - FAQs (17-4321-81)
1 product FAQs found
What is the concentration of Rat IgG2a kappa Isotype Control (eBR2a) (Cat. No. 17-4321-81)?
The concentration is 0.2 mg/mL.