Keratinocyte-SFM (1X), with L-glutamine, 10 x 500 mL - FAQs
Keratinocyte-SFM (1X), with L-glutamine, 10 x 500 mL - FAQs
View additional product information for Keratinocyte-SFM, with L-glutamine - FAQs (17005059)
1 product FAQs found
For how long can I store Keratinocyte-SFM at 4 degrees C?
The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.