beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%
beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%
beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%
Thermo Scientific Chemicals

beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%

CAS: 13035-61-5 | C13H18O9 | 318.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 170120250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13035-61-5
IUPAC Name[3,4,5-tris(acetyloxy)oxolan-2-yl]methyl acetate
Molecular FormulaC13H18O9
InChI KeyIHNHAHWGVLXCCI-UHFFFAOYNA-N
SMILESCC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<1 % (60°C, 2 h) (vacuum)
HPLC>=98.0 %
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only