(1-Ethoxyethylidene)malononitrile, 99+%, Thermo Scientific Chemicals
(1-Ethoxyethylidene)malononitrile, 99+%, Thermo Scientific Chemicals
(1-Ethoxyethylidene)malononitrile, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1-Ethoxyethylidene)malononitrile, 99+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
170291000100 gPlastic bottle
Catalog number 170291000
Price (EUR)
224,00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
224,00
Each
Chemical Identifiers
CAS5417-82-3
IUPAC Name2-(1-ethoxyethylidene)propanedinitrile
Molecular FormulaC7H8N2O
InChI KeyBOSVWXDDFBSSIZ-UHFFFAOYSA-N
SMILESCCOC(C)=C(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder and/or chunks
Infrared spectrumConforms
GC>=99.0 %
Melting point89°C to 93°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only