Hydroxylamine hydrochloride, 97%
Hydroxylamine hydrochloride, 97%
Hydroxylamine hydrochloride, 97%
Thermo Scientific Chemicals

Hydroxylamine hydrochloride, 97%

Hydroxylamine hydrochloride, 97%, Quantity: 1kg, CAS: 5470-11-1, White, Melting Point: 155 deg.C to 158 deg.C, Molecular Formula: H3NO.HCl, Molecular Weight: 69.49, Packaging: Plastic bottle, Percent Purity: 97% | CAS: 5470-11-1 | ClH4NO | 69.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Catalog number 170360010
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Chemical Identifiers
CAS5470-11-1
IUPAC Namehydroxylamine hydrochloride
Molecular FormulaClH4NO
InChI KeyWTDHULULXKLSOZ-UHFFFAOYSA-N
SMILESCl.NO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Melting point151°C to 157°C (Dec)
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only