1-Phenyl-1-cyclohexanecarboxylic acid, 95%
1-Phenyl-1-cyclohexanecarboxylic acid, 95%
1-Phenyl-1-cyclohexanecarboxylic acid, 95%
Thermo Scientific Chemicals

1-Phenyl-1-cyclohexanecarboxylic acid, 95%

CAS: 1135-67-7 | C13H16O2 | 204.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 170750100
Price (USD)
79.65
Online exclusive
88.00
Save 8.35
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
79.65
Online exclusive
88.00
Save 8.35
Each
Chemical Identifiers
CAS1135-67-7
IUPAC Name1-phenylcyclohexane-1-carboxylic acid
Molecular FormulaC13H16O2
InChI KeyQXXHHHWXFHPNOS-UHFFFAOYSA-N
SMILESOC(=O)C1(CCCCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to brown
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Melting point119°C to 124°C
Titration with NaOH94 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only