Soybean Trypsin Inhibitor, powder - FAQs
Soybean Trypsin Inhibitor, powder - FAQs
View additional product information for Soybean Trypsin Inhibitor, powder - FAQs (17075029)
2 product FAQs found
Is Soybean Trypsin Inhibitor a Kunitz-type inhibitor or Bowman-Birk inhibitor?
This product is derived from a Kunitz purification, but in powder form
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What reagents do you offer for cell dissociation, and what are the main differences between them?
Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).
Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.