4-Nitrophenyl chloroformate, 97%
4-Nitrophenyl chloroformate, 97%
4-Nitrophenyl chloroformate, 97%
Thermo Scientific Chemicals

4-Nitrophenyl chloroformate, 97%

CAS: 7693-46-1 | C7H4ClNO4 | 201.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 170805000
Price (USD)
1,513.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
1,513.00
Each
Chemical Identifiers
CAS7693-46-1
IUPAC Name4-nitrophenyl carbonochloridate
Molecular FormulaC7H4ClNO4
InChI KeyNXLNNXIXOYSCMB-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(OC(Cl)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Melting point77°C to 82°C
Titration Argentometric>=96.0 % (ex Cl)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only