Nunc™ Lab-Tek™ Flask on Slide
Thermo Scientific™

Nunc™ Lab-Tek™ Flask on Slide

Grow cell cultures directly on slides with the Thermo Scientific™ Nunc™ Lab-Tek™ Flask on Slide, ideal for single-cell autoradiography and immunofluorescence.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialDescription
170920PolystyreneSlideFlask
177453GlassFlaskette
Catalog number 170920
Price (BRL)
-
Material:
Polystyrene
Description:
SlideFlask
Culture cells directly on microscope slides with the Thermo Scientific™ Nunc™ Lab-Tek™ Flask on Slide, ideal for single-cell autoradiography and single-cell immunofluorescence.

SlideFlask (shown with black cap in photo):

  • Cell culture-treated polystyrene slide is attached to 18 x 50mm flask structure via ultrasonic weld, allowing for long-term, leak-free incubation
  • Upper flask structure snaps away easily after culture, freeing slide for cytostaining and microscopic examination
  • Thermo Scientific™ Nunclon™ Delta certified
  • Coverglass (171862, 18 x 50mm) is available for sample preservation

Flaskette (shown with white cap in phote):

  • Glass slide is sealed with a silicone gasket to a 20 x 50mm upper flask structure
  • Can be sealed to exclude air, useful when embedding cells in resin for electron microscopy
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSlideFlask
IncludesWith black solid cap, non-vented
No. per Case50
No. per Pack5
ColorClear
Culture Area9.0 cm2
MaterialPolystyrene
SterilitySterile
Surface TreatmentCell Culture
Volume (Metric) Working2.5 to 5.0 mL
Wells1
Unit SizeCase of 50