3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrate, 97+%
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrate, 97+%
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrate, 97+%
Thermo Scientific Chemicals

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrate, 97+%

CAS: 1264198-47-1 | C20H12N4Na2O6S2 | 514.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 171010010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1264198-47-1
IUPAC Namedisodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate
Molecular FormulaC20H12N4Na2O6S2
InChI KeyDDIHLFCSEHAOJZ-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=C(N=C(N=N1)C1=CC=CC=N1)C1=CC=C(C=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Suitability(for Cu + Fe determination) Passes test
Appearance (Color)Yellow to yellow-green
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=97.0 % (On dried substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only