6-chloropurine, 99+%
6-chloropurine, 99+%
6-chloropurine, 99+%
Thermo Scientific Chemicals

6-chloropurine, 99+%

CAS: 87-42-3 | C5H3ClN4 | 154.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 171280250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS87-42-3
IUPAC Name6-chloro-7H-purine
Molecular FormulaC5H3ClN4
InChI KeyZKBQDFAWXLTYKS-UHFFFAOYSA-N
SMILESClC1=C2NC=NC2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only