4-Phenyl-3-butyn-2-one, 98%
4-Phenyl-3-butyn-2-one, 98%
4-Phenyl-3-butyn-2-one, 98%
Thermo Scientific Chemicals

4-Phenyl-3-butyn-2-one, 98%

CAS: 1817-57-8 | C10H8O | 144.17 g/mol
製品番号(カタログ番号) 171330050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1817-57-8
IUPAC Name4-phenylbut-3-yn-2-one
Molecular FormulaC10H8O
InChI KeyUPEUQDJSUFHFQP-UHFFFAOYSA-N
SMILESCC(=O)C#CC1=CC=CC=C1
さらに表示
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only