3,4-(Methylenedioxy)aniline, 97%
3,4-(Methylenedioxy)aniline, 97%
3,4-(Methylenedioxy)aniline, 97%
Thermo Scientific Chemicals

3,4-(Methylenedioxy)aniline, 97%

CAS: 14268-66-7 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 171470100
Price (USD)
34.65
Online exclusive
38.70
Save 4.05
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
34.65
Online exclusive
38.70
Save 4.05
Each
Chemical Identifiers
CAS14268-66-7
IUPAC Name2H-1,3-benzodioxol-5-amine
Molecular FormulaC7H7NO2
InChI KeyXGNXYCFREOZBOL-UHFFFAOYSA-N
SMILESNC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to brown to black-brown
Appearance (Form)Crystals and/or chunks or fused solid
Infrared spectrumConforms
Melting point40°C to 46°C
Titration with HClO496.0 to 104.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only