4'-Bromoacetanilide, 98%
4'-Bromoacetanilide, 98%
4'-Bromoacetanilide, 98%
Thermo Scientific Chemicals

4'-Bromoacetanilide, 98%

CAS: 103-88-8 | C8H8BrNO | 214.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 171570250
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.60
Save 5.95
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.60
Save 5.95
Each
Chemical Identifiers
CAS103-88-8
IUPAC NameN-(4-bromophenyl)acetamide
Molecular FormulaC8H8BrNO
InChI KeyMSLICLMCQYQNPK-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to light grey
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point166°C to 170°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only