Bis(4-nitrophenyl) carbonate, 97%
Bis(4-nitrophenyl) carbonate, 97%
Bis(4-nitrophenyl) carbonate, 97%
Thermo Scientific Chemicals

Bis(4-nitrophenyl) carbonate, 97%

CAS: 5070-13-3 | C13H8N2O7 | 304.21 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 171600250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5070-13-3
IUPAC Namebis(4-nitrophenyl) carbonate
Molecular FormulaC13H8N2O7
InChI KeyACBQROXDOHKANW-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(OC(=O)OC2=CC=C(C=C2)[N+]([O-])=O)C=C1
さらに表示
Appearance (Form)Powder
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
Melting point136°C to 144°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only