Dithioerythritol, 99+%
Dithioerythritol, 99+%
Dithioerythritol, 99+%
Thermo Scientific Chemicals

Dithioerythritol, 99+%

CAS: 6892-68-8 | C4H10O2S2 | 154.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 171660010
Price (USD)
64.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
64.00
Each
Chemical Identifiers
CAS6892-68-8
IUPAC Name(2R,3S)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-ZXZARUISSA-N
SMILESO[C@@H](CS)[C@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point82°C to 86°C
Titration Iodimetric>=99.0 %
Solubility(1 % in water) Clear to slightly hazy colorless
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only