3-Methyl-2-buten-1-ol, 99%
3-Methyl-2-buten-1-ol, 99%
3-Methyl-2-buten-1-ol, 99%
Thermo Scientific Chemicals

3-Methyl-2-buten-1-ol, 99%

CAS: 556-82-1 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 171920050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS556-82-1
IUPAC Name3-methylbut-2-en-1-ol
Molecular FormulaC5H10O
InChI KeyASUAYTHWZCLXAN-UHFFFAOYSA-N
SMILESCC(C)=CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4410 to 1.4450 (20°C, 589 nm)
Specific gravity(25°C) 0.857 to 0.867
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only