2-Amino-6-metoxibenzotiazol, 98 %, Thermo Scientific Chemicals
2-Amino-6-metoxibenzotiazol, 98 %, Thermo Scientific Chemicals
2-Amino-6-metoxibenzotiazol, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-6-metoxibenzotiazol, 98 %, Thermo Scientific Chemicals

CAS: 1747-60-0 | C8H8N2OS | 180.23 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
50 g
Envase:
Frasco de vidrio
Número de catálogo 172020050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS1747-60-0
IUPAC Name6-methoxy-1,3-benzothiazol-2-amine
Molecular FormulaC8H8N2OS
InChI KeyKZHGPDSVHSDCMX-UHFFFAOYSA-N
SMILESCOC1=CC=C2N=C(N)SC2=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to yellow to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point165°C to 169°C
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only