Tricine, 98%, for biochemistry
Tricine, 98%, for biochemistry
Tricine, 98%, for biochemistry
Thermo Scientific Chemicals

Tricine, 98%, for biochemistry

CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 172640010
Price (USD)
536.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
536.00
Each
Chemical Identifiers
CAS5704-04-1
IUPAC Name2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}acetic acid
Molecular FormulaC6H13NO5
InChI KeySEQKRHFRPICQDD-UHFFFAOYSA-N
SMILESOCC(CO)(CO)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with HClO4>=97.5 % (On dried basis)
pH4.0 to 6.0 (1 % in water at 20°C)
UVat 260 nm A: =<0.04
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only