Sodium methoxide, pure, anhydrous powder
Sodium methoxide, pure, anhydrous powder
Sodium methoxide, pure, anhydrous powder
Thermo Scientific Chemicals

Sodium methoxide, pure, anhydrous powder

CAS: 124-41-4 | CH3NaO | 54.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 173121000
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.90
Save 6.25
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.90
Save 6.25
Each
Chemical Identifiers
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Alkalinity>=99 % (Total)
Titration>=97 %
Appearance (Color)White to yellow
NaOH + Na2CO3 (as NaOH)=<2 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only