3-Bromotoluene, 98%, Thermo Scientific Chemicals
3-Bromotoluene, 98%, Thermo Scientific Chemicals
3-Bromotoluene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromotoluene, 98%, Thermo Scientific Chemicals

CAS: 591-17-3 | C7H7Br | 171.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
Catalog number 173410100
Price (USD)
33.10
Each
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.10
Each
Chemical Identifiers
CAS591-17-3
IUPAC Name1-bromo-3-methylbenzene
Molecular FormulaC7H7Br
InChI KeyWJIFKOVZNJTSGO-UHFFFAOYSA-N
SMILESCC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to light orange
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5505 to 1.5545 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only