2-Bromo-p-xylene, 97%
2-Bromo-p-xylene, 97%
2-Bromo-p-xylene, 97%
Thermo Scientific Chemicals

2-Bromo-p-xylene, 97%

CAS: 553-94-6 | C8H9Br | 185.06 g/mol
Have Questions?
Catalog NumberQuantity
173421000100 g
Catalog number 173421000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS553-94-6
IUPAC Name2-bromo-1,4-dimethylbenzene
Molecular FormulaC8H9Br
InChI KeyQXISTPDUYKNPLU-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to slightly colored
Appearance (Form)Liquid
Refractive index1.5495 to 1.5515 (20°C, 589 nm)
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only