Phenyl disulfide, 99%
Phenyl disulfide, 99%
Phenyl disulfide, 99%
Thermo Scientific Chemicals

Phenyl disulfide, 99%

CAS: 882-33-7 | C12H10S2 | 218.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
17371050050 gPlastic bottle
Catalog number 173710500
Price (USD)
-
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS882-33-7
IUPAC Name(phenyldisulfanyl)benzene
Molecular FormulaC12H10S2
InChI KeyGUUVPOWQJOLRAS-UHFFFAOYSA-N
SMILESS(SC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point58°C to 62°C
Infrared spectrumConforms
Appearance (Color)White to light yellow to beige
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only