(4-Methoxyphenyl)acetonitrile, 97%
(4-Methoxyphenyl)acetonitrile, 97%
(4-Methoxyphenyl)acetonitrile, 97%
Thermo Scientific Chemicals

(4-Methoxyphenyl)acetonitrile, 97%

CAS: 104-47-2 | C9H9NO | 147.177 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 173760250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104-47-2
IUPAC Name2-(4-methoxyphenyl)acetonitrile
Molecular FormulaC9H9NO
InChI KeyPACGLQCRGWFBJH-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5290 to 1.5330 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only