Pyruvic aldehyde dimethyl acetal, 98%
Pyruvic aldehyde dimethyl acetal, 98%
Pyruvic aldehyde dimethyl acetal, 98%
Thermo Scientific Chemicals

Pyruvic aldehyde dimethyl acetal, 98%

CAS: 6342-56-9 | C5H10O3 | 118.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 173801000
Price (USD)
39.65
Special offer
Online exclusive
Ends: 28-Dec-2026
46.40
Save 6.75
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 28-Dec-2026
46.40
Save 6.75
Each
Chemical Identifiers
CAS6342-56-9
IUPAC Name1,1-dimethoxypropan-2-one
Molecular FormulaC5H10O3
InChI KeyULVSHNOGEVXRDR-UHFFFAOYSA-N
SMILESCOC(OC)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Refractive index1.3970 to 1.3990 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only