1-Iodopropane, 99%, stabilized
1-Iodopropane, 99%, stabilized
1-Iodopropane, 99%, stabilized
Thermo Scientific Chemicals

1-Iodopropane, 99%, stabilized

CAS: 107-08-4 | C3H7I | 169.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 174052500
Price (USD)
105.00
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
105.00
Each
Chemical Identifiers
CAS107-08-4
SpecificationsSpecification SheetSpecification Sheet
GC=<1.5 % (Sum of all by-products)
Refractive index1.5030 to 1.5060 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only