Tetraethylene glycol dimethyl ether, 99%
Tetraethylene glycol dimethyl ether, 99%
Tetraethylene glycol dimethyl ether, 99%
Thermo Scientific Chemicals

Tetraethylene glycol dimethyl ether, 99%

CAS: 143-24-8 | C10H22O5 | 222.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 174112500
Price (USD)
59.60
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
59.60
Each
Chemical Identifiers
CAS143-24-8
IUPAC Name2,5,8,11,14-pentaoxapentadecane
Molecular FormulaC10H22O5
InChI KeyZUHZGEOKBKGPSW-UHFFFAOYSA-N
SMILESCOCCOCCOCCOCCOC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
Refractive index1.4310 to 1.4340 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only