Tiron™
Tiron™
Tiron™
Thermo Scientific Chemicals

Tiron™

CAS: 149-45-1 | C6H10Na2O8S2 | 320.238 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 174140250
Price (USD)
53.90
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.90
Each
Chemical Identifiers
CAS149-45-1
IUPAC Namedisodium 4,5-dihydroxybenzene-1,3-disulfonate
Molecular FormulaC6H4Na2O8S2
InChI KeyISWQCIVKKSOKNN-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC1=CC(=CC(=C1O)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Water4.5 to 6.0 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only