4-Methoxysalicylic acid, 99%
4-Methoxysalicylic acid, 99%
4-Methoxysalicylic acid, 99%
Thermo Scientific Chemicals

4-Methoxysalicylic acid, 99%

CAS: 2237-36-7 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 174240500
Price (USD)
322.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
322.00
Each
Chemical Identifiers
CAS2237-36-7
IUPAC Name2-hydroxy-4-methoxybenzoic acid
Molecular FormulaC8H8O4
InChI KeyMRIXVKKOHPQOFK-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(O)=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Form)Powder
Melting point157°C to 161°C
Appearance (Color)Off-white to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only