4-Ethoxybenzaldehyde, 99%
4-Ethoxybenzaldehyde, 99%
4-Ethoxybenzaldehyde, 99%
Thermo Scientific Chemicals

4-Ethoxybenzaldehyde, 99%

CAS: 10031-82-0 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 174250250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10031-82-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to light brown
AppearanceMay darken on storage
Infrared spectrumConforms
Refractive index1.5574 to 1.5594 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only