Stearic acid, 97%
Stearic acid, 97%
Stearic acid, 97%
Thermo Scientific Chemicals

Stearic acid, 97%

CAS: 57-11-4 | C18H36O2 | 284.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Fibre drum
Catalog number 174490100
Price (USD)
241.65
Online Exclusive
268.00
Save 26.35 (10%)
Each
Quantity:
10 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (USD)
241.65
Online Exclusive
268.00
Save 26.35 (10%)
Each
Chemical Identifiers
CAS57-11-4
IUPAC Nameoctadecanoic acid
Molecular FormulaC18H36O2
InChI KeyQIQXTHQIDYTFRH-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granular powder or granules or flakes
Infrared spectrumConforms
Appearance (Color)White
GC>=96.0 %
Melting point67°C to 71°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only