2,5-Dimethylpyrazine, 99%
2,5-Dimethylpyrazine, 99%
2,5-Dimethylpyrazine, 99%
Thermo Scientific Chemicals

2,5-Dimethylpyrazine, 99%

CAS: 123-32-0 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 174520050
Price (EUR)
50,20
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
50,20
Each
Chemical Identifiers
CAS123-32-0
IUPAC Name2,5-dimethylpyrazine
Molecular FormulaC6H8N2
InChI KeyLCZUOKDVTBMCMX-UHFFFAOYSA-N
SMILESCC1=CN=C(C)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4987 to 1.5007 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only