3-Ethylaniline, 97%
3-Ethylaniline, 97%
3-Ethylaniline, 97%
Thermo Scientific Chemicals

3-Ethylaniline, 97%

CAS: 587-02-0 | C8H11N | 121.18 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
產品號碼 174550500
價格 (TWD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS587-02-0
規格Specification Sheet規格表
GC>=96.0 %
Appearance (Color)Clear colorless to yellow to brown
Refractive index1.5540 to 1.5580 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only