Diethoxydimethylsilane, 97%
Diethoxydimethylsilane, 97%
Diethoxydimethylsilane, 97%
Thermo Scientific Chemicals

Diethoxydimethylsilane, 97%

CAS: 78-62-6 | C6H16O2Si | 148.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 174635000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS78-62-6
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Refractive index1.3805 to 1.3825 (20°C, 589 nm)
Appearance (Color)Clear colorless to slightly colored
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only