Nunclon™ Sphera™ Flasks
Thermo Scientific™
Nunclon™ Sphera™ Flasks
Features a superior cell culture surface with demonstrated performance that elevates stem cell and cell culture research.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture Area
17495125 cm²
17495275 cm²
Catalog number 174951
Price (USD)
421.65
Online exclusive
449.00
Save 27.35
Case of 18
Culture Area:
25 cm²
Price (USD)
421.65
Online exclusive
449.00
Save 27.35
Case of 18
With the super low cell attachment surface, Thermo Scientific™ Nunclon™ Sphera™ Flasks consistently promote 3D spheroids and organoids culture, aiding you in cancer spheroid formation and stem cell differentiation for disease modeling and drug screening.
Optimized performance for 3D cell culture
  • Allows cells to grow in suspension with virtually no cell attachment
  • Supports many different cell types to generate spheroids
  • Promotes consistent formation of cancer spheroids that simulate the 3D structures of tumor growth
  • Demonstrates superior quality for embryoid body formation of pluripotent stem cells with minimal spontaneous differentiation
  • Proven efficacy in supporting long-term organoid culture
Certifications/Compliance
  • Animal origin-free
  • Nonpyrogenic: Endotoxin tested (0.5 EU/mL)
  • Biologically inert: USP biological reactivity class VI
  • High level of sterility: SAL 10-6, ethylene oxide gas sterilization
  • Shelf life: 4 years from the date of manufacture
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAnimal origin-free; nonpyrogenic, endotoxin <0.5 EU/mL; USP biological reactivity class VI; SAL 10-6
Closure TypeFilter
DescriptionT25 Nunclon Sphera EasYFlask, case of 18
No. per Case18
No. per Pack6
Surface CoatingNunclon Sphera
Bottom ShapeFlat
Culture Area25 cm²
SterilitySterile
Unit SizeCase of 18